H-Glu(EDANS)-Pro-Leu-Phe-Ala-Glu-Arg-Lys(DABCYL)-OH
Supplier: Bachem Americas
Certificates
About this item
See also I-1115 and I-1395.
Internally quenched substrate for calpain-1 (µ-calpain) with optimal cleavage motifs flanking the scissile bond. The enzyme showed a more than 18-fold higher turnover rate for the hydrolysis of this FRET substrate based on the amino acid sequence PLFAER than for EVYGMM a sequence derived from the cleavage site of the natural substrate α-spectrin.
Specifications
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD08702758
- Protein/Peptide Name:H-Glu(EDANS)-Pro-Leu-Phe-Ala-Glu-Arg-Lys(DABCYL)-OH
- Molecular Weight:1488.73
- Sequence:H-Glu(EDANS)-Pro-Leu-Phe-Ala-Glu-Arg-Lys(DABCYL)-OH
- Molecular Formula:C₇₂H₉₇N₁₇O₁₆S
Specifications
Frequently Bought Together