Bz-Pro-Phe-Arg-pNA·HCl
Catalog # L-1545.0050BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:50 mg
- Environmentally Preferable:
- Cat. No.:L-1545.0050BA
- Supplier no.:L-1545.0050
Specifications
About this item
:
:679.175 g/mol
Storage Temperature:Freezer
CAS Number:59188-28-2
Formula:C₃₃H₃₉ClN₈O₆
MDL Number:MFCD00039010