Ac-Arg-Leu-Arg-AMC Trifluoroacetate
Supplier: Bachem Americas
Certificates
About this item
Substrates for measuring the chymotryptic, PGPH, and tryptic acitivity of the 20S or 26S proteasome. See also the thrombin substrate I-1140.
Ac-RLR-AMC, fluorogenic substrate for assaying the trypsin-like activity of purified proteasomes (Km = 78 μM).
Specifications
- Source:
- Species:
- Environmentally Preferable:
Specifications
Frequently Bought Together