Boc-Leu-Arg-Arg-AMC · 2 HCl
Catalog # I-1585.0050BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:109358-46-5
- Size:50 mg
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD00133639
- Protein/Peptide Name:Boc-Leu-Arg-Arg-AMC · 2 HCl
- Molecular Weight:773.76
- Sequence:Boc-Leu-Arg-Arg-AMC · 2 HCl
- Molecular Formula:C₃₃H₅₂N₁₀O₇ · 2HCl
- Cat. No.:I-1585.0050BA
- Supplier no.:I-1585.0050
Specifications
About this item
Substrates for measuring the chymotryptic, PGPH, and tryptic acitivity of the 20S or 26S proteasome. See also the thrombin substrate I-1140.
Boc-LLR-AMC is a good substrate for the Kex2 endoprotease (kexin, previously known as proteinase yscF), a membrane-bound, calcium-dependent serine protease from yeast α-cells. Boc-LRR-AMC is also used for determining the trypsin-like activity of the proteasome.