Z-Ala-Arg-Arg-AMC
Catalog # I-1125.0050BA
Supplier: Avantor
CAS Number:
Specifications
- CAS No.:90468-18-1
- Size:50 mg
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD06795827
- Protein/Peptide Name:Z-Ala-Arg-Arg-AMC
- Molecular Weight:692.78
- Sequence:Z-Ala-Arg-Arg-AMC
- Molecular Formula:C₃₃H₄₄N₁₀O₁₇
- Cat. No.:I-1125.0050BA
- Supplier no.:I-1125.0050
Specifications
About this item
Substrates for measuring the chymotryptic, PGPH, and tryptic acitivity of the 20S or 26S proteasome. See also the thrombin substrate I-1140.
Z-ARR-AMC is used for assaying the trypsin-like activity of the 26S proteasome, and cathepsin B.