N(α)-Acetyl-L-tryptophan
Catalog # E-1220.0025BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:25 g
- Cat. No.:E-1220.0025BA
- Supplier no.:E-1220.0025
Specifications
About this item
:
:246.3 g/mol
Storage Temperature:Ambient
CAS Number:1218-34-4
Formula:C₁₃H₁₄N₂O₃
MDL Number:MFCD00065976