Fmoc-Arg(Pbf) 0,3 - 0,6 mmol/g on SASRIN™ resin 200-400 mesh
Catalog # D-1865.0005BA
Supplier: Avantor
CAS Number:
Specifications
- Size:5 g
- Environmentally Preferable:
- Cat. No.:D-1865.0005BA
- Supplier no.:D-1865.0005
Specifications
About this item
:
Storage Temperature:Refrigerator
MDL Number:MFCD00801432