Order Entry
United States
ContactUsLinkComponent
Z-Asn-Gly-OH
Z-Asn-Gly-OH
Catalog # C-4175.0001BA
Supplier:  Bachem Americas
CAS Number:  
Z-Asn-Gly-OH
Catalog # C-4175.0001BA
Supplier:  Bachem Americas
Supplier Number:  4039578.0001
CAS Number:  

Specifications

  • CAS No.:
    56675-97-9
  • Source:
  • Species:
  • Size:
    1 g
  • Environmentally Preferable:
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD08458674
  • Protein/Peptide Name:
    Z-Asn-Gly-OH
  • Molecular Weight:
    323.31
  • Sequence:
    Z-Asn-Gly-OH
  • Molecular Formula:
    C₁₄H₁₇N₃O₆
  • Cat. No.:
    C-4175.0001BA
  • Supplier no.:
    C-4175.0001

Specifications

About this item