Z-Phe-Tyr(tBu)-Diazomethylketone
Supplier: Bachem Americas
Certificates
About this item
Please see also the inhibitors H-7650, N-1000, N-1040, N-1105, N-1125, N-1315, N-1320, N-1380, N-1395, and N-1855.
Irreversible cathepsin L inhibitor which is ca 10'000 times more effective in inactivating cathepsin L than cathepsin S.
Specifications
- CAS No.:114014-15-2
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD00672349
- Protein/Peptide Name:Z-Phe-Tyr(tBu)-diazomethylketone
- Molecular Weight:542.63
- Sequence:Z-Phe-Tyr(tBu)-diazomethylketone
- Molecular Formula:C₃₁H₃₄N₄O₅
Specifications
Frequently Bought Together