Fmoc-Pro-Pro-Pro-OH
Supplier: Avantor
Certificates
About this item
For Fmoc-dipeptides Fmoc-Xaa-Gly-OH and Fmoc-Xaa-Pro-OH, which can be used as building blocks in Fmoc-SPPS, please see also the subfamily 'Fmoc-Dipeptide Building Blocks' in the 'Amino Acid Derivatives' section. Pseudoproline dipeptides and isoacyl dipeptides can be found in this section as well. In case of a strong tendency towards diketopiperazine formation during Fmoc-SPPS, Fmoc-dipeptides have been coupled even at the risk of a some epimerization.
Fmoc-triproline, potential building block for chiral stationary phases.
Specifications
- CAS No.:134303-96-1
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD08458580
- Protein/Peptide Name:Fmoc-Pro-Pro-Pro-OH
- Molecular Weight:531.61
- Sequence:Fmoc-Pro-Pro-Pro-OH
- Molecular Formula:C₃₀H₃₃N₃O₆
Specifications
Frequently Bought Together