Fmoc-Pro-Pro-Pro-Pro-Pro-OH
Supplier: Bachem Americas
Certificates
About this item
For Fmoc-dipeptides Fmoc-Xaa-Gly-OH and Fmoc-Xaa-Pro-OH, which can be used as building blocks in Fmoc-SPPS, please see also the subfamily 'Fmoc-Dipeptide Building Blocks' in the 'Amino Acid Derivatives' section. Pseudoproline dipeptides and isoacyl dipeptides can be found in this section as well. In case of a strong tendency towards diketopiperazine formation during Fmoc-SPPS, Fmoc-dipeptides have been coupled even at the risk of a some epimerization.
Fmoc-pentaproline, Fmoc-PPPPP, potential building block for polyproline-based chiral stationary phases.
Specifications
- CAS No.:454693-94-8
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD08458582
- Protein/Peptide Name:Fmoc-Pro-Pro-Pro-Pro-Pro-OH
- Molecular Weight:725.84
- Sequence:Fmoc-Pro-Pro-Pro-Pro-Pro-OH
- Molecular Formula:C₄₀H₄₇N₅O₈
Specifications
Frequently Bought Together