Order Entry
United States
Orders LinkContactUsLinkComponent
Fmoc-Phe-Phe-OH

Fmoc-Phe-Phe-OH

Supplier:   Bachem Americas
Fmoc-Phe-Phe-OH
Supplier:  Bachem Americas
Certificates

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

For Fmoc-dipeptides Fmoc-Xaa-Gly-OH and Fmoc-Xaa-Pro-OH, which can be used as building blocks in Fmoc-SPPS, please see also the subfamily 'Fmoc-Dipeptide Building Blocks' in the 'Amino Acid Derivatives' section. Pseudoproline dipeptides and isoacyl dipeptides can be found in this section as well. In case of a strong tendency towards diketopiperazine formation during Fmoc-SPPS, Fmoc-dipeptides have been coupled even at the risk of a some epimerization.

The nanotube formation of Fmoc-FF has been studied by Gazit and Reches. Jayawarna et al. found that this self-assembling dipeptide forms fibrous hydrogels at pH<8. Such hydrogels are excellently suited for 3D cell microculture formation, they can be modified by addition of Fmoc-amino acids as Fmoc-Lys-OH (B-1870).

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • B-2150.0001BA
  • B-2150.0005BA
  • Protein/Peptide Name:
  • Fmoc-Phe-Phe-OH
  • Fmoc-Phe-Phe-OH
  • CAS No.:
  • 84889-09-8
  • 84889-09-8
  • MDL No.:
  • MFCD00237669
  • MFCD00237669
  • Sequence:
  • Fmoc-Phe-Phe-OH
  • Fmoc-Phe-Phe-OH
  • Molecular Weight:
  • 534.61
  • 534.61
  • Molecular Formula:
  • C₃₃H₃₀N₂O₅
  • C₃₃H₃₀N₂O₅
  • Storage Conditions:
  • –20 ± 5 °C
  • –20 ± 5 °C

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $355.56
    $355.56
    Each (1g)
       $355.56
    Minimum order quantity is 1

    Supplier Number: 4015688.0001
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    B-2150.0001BA
    Protein/Peptide Name
    Fmoc-Phe-Phe-OH
    CAS No.
    84889-09-8
    MDL No.
    MFCD00237669
    Sequence
    Fmoc-Phe-Phe-OH
    Molecular Weight
    534.61
    Molecular Formula
    C₃₃H₃₀N₂O₅
    Storage Conditions
    –20 ± 5 °C
  • 5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $1,349.82
    $1,349.82
    Each (5g)
       $1,349.82
    Minimum order quantity is 1

    Supplier Number: 4015688.0005
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    B-2150.0005BA
    Protein/Peptide Name
    Fmoc-Phe-Phe-OH
    CAS No.
    84889-09-8
    MDL No.
    MFCD00237669
    Sequence
    Fmoc-Phe-Phe-OH
    Molecular Weight
    534.61
    Molecular Formula
    C₃₃H₃₀N₂O₅
    Storage Conditions
    –20 ± 5 °C