N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine pentafluorophenyl ester
Catalog # B-1100.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:5 g
- Cat. No.:B-1100.0005BA
- Supplier no.:B-1100.0005
Specifications
About this item
:
:477.39 g/mol
Storage Temperature:Freezer
CAS Number:86060-86-8
Formula:C₂₄H₁₆F₅NO₄
MDL Number:MFCD00065612