Order Entry
United States
Orders LinkContactUsLinkComponent
Boc-Trp-OMe

Boc-Trp-OMe

Boc-Trp-OMe
Supplier:  
Certificates

0 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Sequence: Boc-Trp-OMe

Synonyms:  N-Boc-L-tryptophan methyl ester
  • CAS Number:33900-28-6
  • Formula:C₁₇H₂₂N₂O₄
  • MDL Number:MFCD01075094
  • Molecular Weight:318.373 g/mol
  • Storage Temperature:Freezer
Product Family Options