Nɑ-(tert-Butoxycarbonyl)-N-ε-carbobenzoxy-L-lysine
Supplier: Bachem Americas
Certificates
About this item
CAS Number:2389-45-9
:
Formula:C₁₉H₂₈N₂O₆
MDL Number:MFCD00065584
:380.44 g/mol
Storage Temperature:Ambient
EINECS:219-221-0
Frequently Bought Together