Chloride Low Range Test Kit, Model 8-P, Hach
Catalog # 89231-234
Supplier: Avantor
Product available on GSA Advantage®
CAS Number:
Specifications
- Chemical method:Silver nitrate
- Description:Chloride Low Range Test Kit, Model 8-P
- Model:8-P
- Method:Drop count titration/Silver nitrate
- Case Style:B
- Increments:Smallest steps - 20
- Cat. No.:89231-234
- Range:5 - 100 mg/l Cl-, 20 - 400 mg/l Cl-
- Supplier no.:144001
- No. of tests:100
Specifications
About this item
Using silver nitrate titrant, this kit provides fast, accurate detection of chloride.
- Low cost and easy to use drop count titration method using a silver nitrate titrant
- Ideal for quickly determining quantitative results
*mg/L unless otherwise noted, ppb = μg/L, ppm = mg/L.; gpg = grains per gallon; 1 gpg = 17.1 mg/L or 17.1 ppm. 1EPA-accepted for wastewater. Check with your regulatory agency.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.