Order Entry
United States
ContactUsLinkComponent
TPH in Water, Test Kit, Hach
TPH in Water, Test Kit, Hach
  97027-084
 :  Avantor
 :  
undefined
TPH in Water, Test Kit, Hach
  97027-084
 :  Avantor
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Description:
    TPH in Water, Test Kit
  • Model:
    PocketColorimeter™ II
  • Case Style:
    Q
  • Wavelength Accuracy:
    Fixed wavelength ± 2 nm varies with model
  • Compliance:
    European CE Mark, IP 67 ( enclosure)
  • Display:
    LCD, backlit
  • Light Source:
    Light Emitting Diode (LED)
  • Optics:
    Detector Silicon
  • W×D×H:
    155×61×35 mm
  • Cat. No.:
    97027-084
  • Range:
    2, 5, 10, 20 ppm threshold
  • Supplier no.:
    2774200
  • No. of tests:
    Up to 18

 

 

For personnel responsible for underground storage tanks, off-site waste impounds, petrochemical plants, and waste remediation sites, Hach TPH kits provide fast accurate results at low levels.

  • Provides fast, accurate results at low levels
  • Pocket Colorimeter™ II is waterproof, IP67 rated, and datalogs last 10 points with measurement time
  • Used by personnel responsible for underground storage tanks, offsite waste impounts, petrochemical plants, and waste remediate sites

This water kit has thresholds of 2, 5, 10, and 20 ppm.

 : 2 yr warranty.

Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.