BART Test for Blue-Green Algae, Hach
97026-616
: Avantor
Product available on GSA Advantage®
:
- Description:BART Test for Blue-Green Algae, Hach
- Environmentally Preferable:
- Cat. No.:97026-616
- Supplier no.:2432709
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria.
- Simple yet effective method for monitoring the population size and/or activity of specific groups of bacteria.
- Easy to use, requiring no elaborate or costly equipment and no specialized training.
- Effective and affordable tests are easy to interpret and can be performed at room temperature in virtually any environment.
: 9/pk.