Order Entry
United States
ContactUsLinkComponent
Nitrogen, Nitrate Color Disc Test Kit, Model NI-14, Hach
Nitrogen, Nitrate Color Disc Test Kit, Model NI-14, Hach
Catalog # 97026-822
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  
undefined
Nitrogen, Nitrate Color Disc Test Kit, Model NI-14, Hach
Catalog # 97026-822
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Chemical method:
    4AAP (4-Aminoantipyrine)
  • Description:
    Nitrogen, Nitrate Color Disc Test Kit, Model NI-14
  • Environmentally Preferable:
  • Model:
    NI-14
  • Method:
    Color disc/ Cadmium reduction
  • Case Style:
    B
  • Increments:
    0.2
  • Cat. No.:
    97026-822
  • Range:
    0.02 - 1 mg/l NO₃--N, 0.2 - 10 mg/l NO₃--N
  • Supplier no.:
    1416100
  • No. of tests:
    50

Specifications

About this item

Utilizing the simple color disc method, this kit measures Nitrogen, Nitrate as NO3 and contains 50 tests.

  • Simple, lowcost color disc method is the most accurate visual test kit method
  • Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field