ContactUsLinkComponent
Optical Sealing Tape, Bio-Rad
Optical Sealing Tape, Bio-Rad
Catalog # 101645-688
Supplier:  Bio-Rad
undefined
Optical Sealing Tape, Bio-Rad
Catalog # 101645-688
Supplier:  Bio-Rad
Supplier Number:  2239444
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Optical Sealing Tape
  • Cat. No.:
    101645-688
  • Supplier no.:
    2239444
Specifications
About this item
Optical sealing tape is an adhesive-based sealing option for optical assays demanding a low-fluorescent background.
The tape is validated for real-time PCR on iCycler iQ systems (Bio-Rad) and for melt-curve analysis on the LightScanner system (Idaho Technology, Inc.). The moderate-strength adhesive layer ensures easy removal of the tape after reactions. Microseal 'B' adhesive seals are recommended if strong sealing performance is needed.
Ordering information: Package of 100.