Dissolved Oxygen AccuVac® Kit, Hach
Catalog # 89231-116
Supplier: Avantor
Product available on GSA Advantage®
CAS Number:
Specifications
- Chemical method:HRDO method
- Description:Dissolved Oxygen AccuVac® Kit
- Model:AccuVac® Kit
- Method:Color disc/HRDO
- Case Style:G
- Cat. No.:89231-116
- Range:0 - 15 mg/l O₂
- Supplier no.:2515050
- No. of tests:25
Specifications
About this item
Low DO levels usually indicate serious pollution.
- Simple, lowcost color disc method is the most accurate visual test kit method
- Achieve fast results with an easytocarry test kit
Adequate amounts are crucial for fish life, but conversely, DO must be excluded from boiler feed water to prevent corrosion. The simple, low-cost color disc method is the most accurate visual test kit method.