Chlorine (Free and Total) Test Kit, Model CN-80, Hach
Catalog # 89231-330
Supplier: Avantor
Product available on GSA Advantage®
CAS Number:
Specifications
- Chemical method:DPD (N,N-diethyl-p-phenylenediamine)
- Description:Chlorine (Free and Total) Test Kit, Model CN-80
- Model:CN-80
- Method:Color Disc/DPD
- Case Style:D
- Cat. No.:89231-330
- Range:0 - 3.4 mg/l, 0.5 - 10 mg/l, 0.02 - 0.7 mg/l
- Supplier no.:2129000
- No. of tests:100
Specifications
About this item
Simple, low-cost color disc method is the most accurate visual test kit method.
- Simple, lowcost color disc method is the most accurate visual test kit method
- Achieve fast results with an easytocarry test kit
*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.