Specifications
- Description:Soil and Irrigation Water Test Kit, Model SIW-1
- Environmentally Preferable:
- Cat. No.:89231-410
- Supplier no.:2496000
Specifications
About this item
A test kit for professional scientists and non-scientists alike, the SIW-1 Soil and Irrigation Water Test Kit is ideal for fertility analysis, site characterization, and irrigation water quality assessment under a wide variety of conditions.
- Uses color disc, drop count titration, and dipstick method
- Includes manual for interpretation of Soil and Irrigation Water test results
Farmers, researchers, extension agents, and educators can use the versatile test kit at remote sites, in the classroom, or even in areas lacking electricity.