Order Entry
United States
ContactUsLinkComponent
Water Quality Meter pH Kit, Sper Scientific
Water Quality Meter pH Kit, Sper Scientific
  75997-560
undefined
Water Quality Meter pH Kit, Sper Scientific
  75997-560
 :  850086

 

  • Description:
    Water Quality Meter pH Kit
  • Dimensions:
    280 × 190 × 63 mm (11 × 7.5 × 2.5")
  • Weight:
    1.1 kg (2.5 lbs.)
  • Cat. No.:
    75997-560

 

 

Interchangeable plug and play probes let this versatile meter read pH, mV, ORP, conductivity, salinity, TDS, DO and temperature.

  • Automatic calibration recognition of pH buffers 4, 7 and 10
  • Auto power off, hold, min/max
  • Fold-out easel back and tripod screw

Order probes only for parameters currently required, and add additional probes in the future. The back-lit display simultaneously reads temperature in user selectable °C & °F. All measurements are automatically temperature compensated. DO measurements can be compensated for salinity, while Oxygen in Air (O2) can be compensated for altitude.

Comes with 9V battery and hard-shell, foam-lined carrying case with room for one or two probes.

Kit Dimensions: 11" × 9½" × 2" (279 × 241 × 51 mm)

 : Water Quality Meter pH Kit includes everything needed for immediate field use as a pH meter:

Water Quality Meter #850081
pH Probe #840016
ATC Temperature Probe #840038
pH buffers 4, 7 & 10
De-ionized water
9V battery
Instructions
Hard Shell Foam-lined Carrying Case