Order Entry
United States
ContactUsLinkComponent
Acetate Buffer Solution, pH 4.0, 100 mL, Hach
Acetate Buffer Solution, pH 4.0, 100 mL, Hach
Catalog # 97026-592
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  
undefined
Acetate Buffer Solution, pH 4.0, 100 mL, Hach
Catalog # 97026-592
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Size:
    100 mL
  • pH buffer type:
    Acetate
  • Cat. No.:
    97026-592
  • pH:
    4.0
  • Supplier no.:
    1490932
  • No. of tests:
    100

Specifications

About this item

Acetate Buffer Solution, pH 4.0

For use with AutoCAT™ 9000 titrator for Total and/or Free Chlorine. Method: Amperometric.