Order Entry
United States
ContactUsLinkComponent
Hardness, Iron, and pH Test Kit, Model HA-62A, Hach
Hardness, Iron, and pH Test Kit, Model HA-62A, Hach
Catalog # 89231-248
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  
undefined
Hardness, Iron, and pH Test Kit, Model HA-62A, Hach
Catalog # 89231-248
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Description:
    Hardness, Iron, and pH Test Kit, Model HA-62A
  • Cat. No.:
    89231-248
  • Range:
    pH 6.6 - 8.4
  • Supplier no.:
    183701
  • No. of tests:
    100

Specifications

About this item

A multi-parameter test kit for hardness, iron & pH, this kit provides an easy-to-use, cost effective way to determine the quality of your water.

  • Popular tests at a great price in Hach's convenient and protective test kit carrying case.
  • Uses color disc and drop count titration methods
  • Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.