Environmental Quality Meter, Sper Scientific
14223-290
- Description:Environmental Quality Meter
- Cat. No.:14223-290
- Supplier no.:850071
Using interchangeable probes (sold separately) this versatile environmental meter reads airspeed, air temperature, RH, light and thermocouple and IR temperature
You can order probes for only the parameters currently required and add additional probes in the future. Intelligent probes electronically retain calibration information. No recalibration is required when changing probes. The display simultaneously reads temperature in user selectable ºC and ºF and all measurements are automatically temperature compensated. The RS232 port enables communication with a computer. The environmental quality meter also features automatic shut off, hold and min/max, has a fold out easel back and a tripod screw.
: Comes with a 9V battery and a hard-shell foam-lined carrying case with room for one or two probes.