Hydrogen Sulfide Test Kit, Model HS-C, Hach
Catalog # 89231-120
Supplier: Avantor
Product available on GSA Advantage®
CAS Number:
Specifications
- Chemical method:Effervescence of H2S
- Description:Hydrogen Sulfide Test Kit, Model HS-C
- Model:HS-C
- Method:Color chart/ Effervescence of H2S
- Case Style:M
- Increments:Steps: 0.0, 0.1, 0.3, 0.5, 0.7, 1.0, 2.0, 5.0
- Cat. No.:89231-120
- Range:0 - 5 mg/l H₂S
- Supplier no.:2537800
- No. of tests:18
Specifications
About this item
Resulting from the anaerobic decomposition of organic matter, hydrogen sulfide is present in many water supplies.
- Most economical method for determining hydrogen sulfide content in water samples
- Uses reliable, cost effective color comparison method
- Treated reagent papers are supplied for 100 tests; effervescence tablets are supplied for 18 tests
Sewage and industrial wastes are other sources of sulfide pollution. Highly toxic, this compound has a characteristic rotten egg odor, which can be detected long before harmful concentrations are reached. In the HS-C kit, sulfide discolors a detection paper. Results are obtained by comparison of the color to a printed chart.