ContactUsLinkComponent
Ward's® Gram Staining and Bacterial Morphology Lab Activity
Ward's® Gram Staining and Bacterial Morphology Lab Activity
Catalog # 470190-710
Supplier:  Avantor
undefined
Ward's® Gram Staining and Bacterial Morphology Lab Activity
Catalog # 470190-710
Supplier:  Avantor
Specifications
  • Description:
    Staining and Morphology Lab Activity
  • No. of lab groups:
    4
  • Cat. No.:
    470190-710
Specifications
About this item
In this lab, students perform a Gram stain on various species of bacteria.
  • Kit Aligned with National Standards
  • Lab Employs Framework Science Standard LS1
  • Students Learn to Recognize Gram-Positive and Gram-Negative Bacteria
The results, after observation through a light microscope, allow each sample to be designated either Gram negative or Gram positive.
Students then describe the unique properties of each type of bacterium's cell wall and the mechanism through which the bacteria are differentiated during the procedure.
Canada: Containment Level 2 – PHAC License required.
Ordering information: This lab contains 1 box of pre-cleaned microscope slides, 1 coupon redeemable for five microorganism cultures and additional media, 20 sterile culture swabs, a teacher's guide, students' guides, and stains and decolorizers for a gram stain test. A microscope with an oil-immersion objective, oil for oil immersion objective, clothespins, burners, paper towels, and distilled water are needed but not included.
Delivery: The kit will ship separately from the live component. Kits do not contain perishable components and generally ship via UPS ground. Perishable components will ship via overnight or second day delivery.