ContactUsLinkComponent
Ward's® Paternity Testing Lab Activity
Ward's® Paternity Testing Lab Activity
Catalog # 470190-634
Supplier:  Avantor
undefined
Ward's® Paternity Testing Lab Activity
Catalog # 470190-634
Supplier:  Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    WARD'S Paternity Testing Kit
  • No. of lab groups:
    8
  • Cat. No.:
    470190-634
Specifications
About this item
How does DNA fingerprinting determine paternity?
  • Kit employs NGSS Life Science DCIs LS1 and LS3
  • Pre- and post-laboratory assessments included
  • Materials for eight lab groups
  • Assessments and vocabulary words target Common Core standards
  • Kit aligns with National Standards
With this lab, students gain experience using agarose gel and analyzing results. Students use the results to isolate the genes from a child that were not inherited from his or her mother and determine who the father is. The students' understanding of this exercise helps them gain experience analyzing data and autoradiograms. This kit employs NGSS Life Science Disciplinary Core Ideas (LS1) and (LS3).
This kit is aligned with all published National Standards. Pre and post-laboratory assessments and vocabulary words target specific Science and Engineering Practices and Common Core standards. The teacher guide also contains specific "Did You Know" concepts that link the real world with the scientific standards explored. A student copymaster is also included.
Ordering information: This kit includes 200 mL agarose, 0.8%; 500 mL 5X TBE running buffer; one bottle of Ward’s QUICKView DNA stain concentrate, 60 mL; 80 gel-staining trays; coupon redeemable for the following DNA samples: 80 µL each of "Mother"; "Child"; "Alleged father #1"; "Alleged father #2".
Agarose electrophoresis chambers, power supplies, a water bath or microwave, micropipets with tips, gloves apron, goggles, lab markers, and distilled water are required but not included.