Order Entry
United States
ContactUsLinkComponent
pH SD Card Logger Kit, Sper Scientific
pH SD Card Logger Kit, Sper Scientific
Catalog # 89168-702
Supplier:  Sper Scientific
undefined
pH SD Card Logger Kit, Sper Scientific
Catalog # 89168-702
Supplier:  Sper Scientific
Supplier Number:  850061

Specifications

  • Resolution:
    pH: 0.01
    mV: 1
  • Interface:
    RS232
  • Description:
    pH SD card logger kit
  • W×D×H:
    279×241×51 mm (11 ×9.5×2")
  • Cat. No.:
    89168-702
  • Accuracy:
    pH: ±0.04
    RDG: ±0.5%+2 d
  • Range:
    pH: 0 - 14
    mV: –1999 - +1999
    ORP: –0 - +1999 mV
  • Supplier no.:
    850061

Specifications

About this item

The pH SD card logger reads pH, ORP, and temperature, and records data onto standard SD memory cards (like a digital camera)

The pH SD card logger kit includes everything necessary for immediate field use. Each order includes pH SD card logger, sealed epoxy body reference combination electrode with a BNC connector and a 2½' cable, ATC temperature probe, pH buffers 4, 7, 10 and deionized water, batteries, and instructions. All components are contained in a slim but protective hard-shell, foam-lined case.

When full, simply exchange SD cards for unlimited data storage. The removable SD card is easy to transport and insert into a computer’s memory card slot or SD card reader. The device uploads preformatted data to Excel, without the need for additional software and cables. Additional SD cards are readily available in a variety of memory sizes. They can also stream data directly to a computer using the optional software and cables. Each data set includes pH, temperature, time, and date. The device features ATC (automatic temperature compensation) or manual temperature adjustment, automatic buffer recognition, min./max., auto power off, low battery indicator, and hold function. This well-made unit is extremely accurate with fast response times, and includes a fold-out easel back and a large easy-to-read backlit LCD.

Ordering information: Each pH SD card logger kit includes a 5-year warranty.