Order Entry
United States
ContactUsLinkComponent
 
pH SD Card Logger, Sper Scientific
  89168-700
undefined
pH SD Card Logger, Sper Scientific
  89168-700
 :  850060

 

  • Resolution:
    pH: 0.01
    mV: 1
  • Description:
    pH SD card logger (meter only)
  • Weight:
    0.45 kg
  • W×D×H:
    178×70×44 mm (7×2.75×1.75")
  • Cat. No.:
    89168-700
  • Accuracy:
    pH: ±0.04
    RDG: ±0.5%+2 d
  • Range:
    pH: 0 - 14
    mV: –1999 - +1999
    ORP: –0 - +1999 mV
  • Supplier no.:
    850060

 

 

The ph SD Card Logger reads pH, ORP, and temperature

Note: Probe not included. ATC temperature probe available as optional accessory. Unit will accept any pH or ORP probe with a BNC connector.

The unit records data onto standard SD memory cards (like a digital camera). When full, simply exchange SD cards for unlimited data storage. The removable SD card is easy to transport and insert into a computer’s memory card slot or SD card reader. It uploads preformatted data to Excel, without the need for additional software and cables. Additional SD cards are readily available in a variety of memory sizes. The unit can also stream data directly to a computer using the optional software and cables. Each data set includes pH, temperature, time, and date. It features ATC (automatic temperature compensation) or manual temperature adjustment, automatic buffer recognition, min./max., auto power off, low battery indicator, and hold function. This well-made unit is extremely accurate with fast response times, and includes a fold-out easel back and a large easy-to-read backlit LCD.

 : Supplied ready to use in a soft carrying case with 6 AA batteries, instruction manual, and a 2 GB SD card. Each unit includes a 5-year warranty.