Order Entry
United States
ContactUsLinkComponent
 
(d(CH₂)₅¹,Tyr(Me)²,Thr⁴,Orn⁸,Tyr-NH₂⁹)-Vasotocin Trifluoroacetate
 
(d(CH₂)₅¹,Tyr(Me)²,Thr⁴,Orn⁸,Tyr-NH₂⁹)-Vasotocin Trifluoroacetate

 

  • CAS No.:
    114056-26-7
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD18910832
  • Protein Synonyms:
    OVTA
  • Protein/Peptide Name:
    (d(CH₂)₅¹,Tyr(Me)²,Thr⁴,Orn⁸,Tyr-NH₂⁹)-Vasotocin Trifluoroacetate
  • Molecular Weight:
    1154.42
  • Sequence:
    β-Mercapto-β,β-cyclopentamethylene-propionyl-Tyr(Me)-Ile-Thr-Asn-Cys-Pro-Orn-Tyr-NH₂
  • Molecular Formula:
    C₅₄H₇₉N₁₁O₁₃S₂

 

Frequently Bought Together