Fibrinogen-Binding Inhibitor Peptide
H-9140.0005BA
:
- CAS No.:89105-94-2
- Size:5 mg
- Storage Conditions:–20 ± 5 °C
- Protein/Peptide Name:Fibrinogen-Binding Inhibitor Peptide
- Molecular Weight:1189.30
- Sequence:H-His-His-Leu-Gly-Gly-Ala-Lys-Gln-Ala-Gly-Asp-Val-OH
- Molecular Formula:C₅₀H₈₀N₁₈O₁₆
- Cat. No.:H-9140.0005BA
- Supplier no.:H-9140.0005
The synthetic dodecapeptide HHLGGAKQAGDV represents the specific platelet receptor recognition site of the human fibrinogen γ chain (residues 400-411). It is a potent inhibitor of the binding of fibrinogen, fibronectin, and von Willebrand factor to thrombin- or ADP-stimulated platelets. The peptide has also been used in studying phosphorylation processes associated with platelet activation.