Order Entry
United States
ContactUsLinkComponent
Acetyl-alpha-MSH (11-13)
Acetyl-alpha-MSH (11-13)
Supplier:  Bachem Americas
Certificates
Acetyl-alpha-MSH (11-13)
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    57899-96-4
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD18782493
  • Protein/Peptide Name:
    Acetyl-alpha-MSH (11-13)
  • Molecular Weight:
    383.49
  • Sequence:
    Ac-Lys-Pro-Val-NH₂
  • Molecular Formula:
    C₁₈H₃₃N₅O₄

Specifications

Frequently Bought Together