Order Entry
United States
ContactUsLinkComponent
Human TRAP-14 Trifluoroacetate
Human TRAP-14 Trifluoroacetate
Supplier:  Bachem Americas
Certificates
Human TRAP-14 Trifluoroacetate
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    137339-65-2
  • Species:
    Human
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD18782486
  • Protein Synonyms:
    SFLLRNPNDKYEPF|Proteinase Activated Receptor 1 (1-14)|Coagulation Factor II Receptor (1-14)|PAR-1 (1-14)|Thrombin Receptor (1-14)|Thrombin Receptor Activator Peptide 14
  • Protein/Peptide Name:
    TRAP-14 Trifluoroacetate
  • Molecular Weight:
    1739.95
  • Sequence:
    Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe
  • Molecular Formula:
    C₈₁H₁₁₈N₂₀O₂₃

Specifications

Frequently Bought Together