Order Entry
United States
ContactUsLinkComponent
Trypanothione
Trypanothione
Supplier:  Bachem Americas
Certificates
Trypanothione
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    96304-42-6
  • Storage Conditions:
    –20 ± 5 °C
  • Protein/Peptide Name:
    Trypanothione
  • Molecular Weight:
    721.86
  • Sequence:
    Trypanothione was purified from the insect trypanosomatid Crithidia fasciculata and identified as a covalently linked adduct of glutathione and spermidine. It is utilized as a substrate by trypanothione reductase, a unique enzyme found in trypanosomes and leishmanias (Km = 51 µM).
  • Molecular Formula:
    C₂₇H₄₇N₉O₁₀S₂

Specifications

Frequently Bought Together