H(-Asn-Pro-Asn-Ala)₆-OH
Catalog # H-6340.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:114332-71-7
- Size:1 mg
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD00237687
- Protein/Peptide Name:H(-Asn-Pro-Asn-Ala)₆-OH
- Molecular Weight:2396.44
- Sequence:H(-Asn-Pro-Asn-Ala)₆-OH
- Molecular Formula:C₉₆H₁₄₆N₃₆O₃₇
- Cat. No.:H-6340.0001BA
- Supplier no.:H-6340.0001
Specifications
About this item
Antibodies to these peptides from the repeating region of the CS protein recognized native CS protein and blocked Plasmodium falciparum sporozoite invasion of human hepatoma cells in vitro.
- Hexamer of NPNA.