FLAG Epitope Peptide Trifluoroacetate
Supplier: Bachem Americas
Certificates
About this item
The FLAG Epitope Peptide consists of eight amino acids (DYKDDDDK) and represents a versatile fusion tag for the purification of recombinant proteins. It can be enzymatically removed with enterokinase which cleaves C-terminally to the tag sequence. The peptide is commonly used for the elution of immunoaffinity chromatography-purified recombinant proteins.
Specifications
- Source:
- Species:
- Environmentally Preferable:
Specifications
Frequently Bought Together