Order Entry
United States
ContactUsLinkComponent
epsilonAhx-omega-Conotoxin GVIA Trifluoroacetate,Biotin
epsilonAhx-omega-Conotoxin GVIA Trifluoroacetate,Biotin
Supplier:  Bachem Americas
Certificates
epsilonAhx-omega-Conotoxin GVIA Trifluoroacetate,Biotin
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    151928-23-3
  • Conjugation:
    Biotin
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD18782359
  • Protein/Peptide Name:
    epsilonAhx-omega-Conotoxin GVIA Trifluoroacetate
  • Molecular Weight:
    3376.85
  • Sequence:
    Biotinyl-εAhx-Cys-Lys-Ser-Hyp-Gly-Ser-Ser-Cys-Ser-Hyp-Thr-Ser-Tyr-Asn-Cys-Cys-Arg-Ser-Cys-Asn-Hyp-Tyr-Thr-Lys-Arg-Cys-Tyr-NH₂
  • Molecular Formula:
    C₁₃₆H₂₀₇N₄₁O₄₆S₇

Specifications

Frequently Bought Together