Order Entry
United States
ContactUsLinkComponent
H-p-Chloro-Phe-D-Cys-beta-(3-pyridyl)-Ala-D-Trp-Lys-tBu-Gly-Cys-2-Nal-NH₂ Trifluoroacetate
H-p-Chloro-Phe-D-Cys-beta-(3-pyridyl)-Ala-D-Trp-Lys-tBu-Gly-Cys-2-Nal-NH₂ Trifluoroacetate
Supplier:  Bachem Americas
Certificates
H-p-Chloro-Phe-D-Cys-beta-(3-pyridyl)-Ala-D-Trp-Lys-tBu-Gly-Cys-2-Nal-NH₂ Trifluoroacetate
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    209006-18-8
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD18633553
  • Protein Synonyms:
    PRL-2915
  • Protein/Peptide Name:
    H-p-Chloro-Phe-D-Cys-beta-(3-pyridyl)-Ala-D-Trp-Lys-tBu-Gly-Cys-2-Nal-NH₂ Trifluoroacetate
  • Molecular Weight:
    1175.87
  • Sequence:
    H-p-Chloro-Phe-D-Cys-β-(3-pyridyl)-Ala-D-Trp-Lys-tBu-Gly-Cys-2-Nal-NH₂
  • Molecular Formula:
    C₅₉H₇₁ClN₁₂O₈S₂

Specifications

Frequently Bought Together