Order Entry
United States
ContactUsLinkComponent
H-Tyr-Tyr-Tyr-OH
H-Tyr-Tyr-Tyr-OH
Supplier:  Bachem Americas
Certificates
H-Tyr-Tyr-Tyr-OH
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    7390-78-5
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00055745
  • Protein Synonyms:
    YYY
  • Protein/Peptide Name:
    H-Tyr-Tyr-Tyr-OH
  • Molecular Weight:
    507.54
  • Sequence:
    H-Tyr-Tyr-Tyr-OH
  • Molecular Formula:
    C₂₇H₂₉N₃O₇

Specifications

Frequently Bought Together