Mouse;Rat alpha-CGRP (8-37) Trifluoroacetate
Supplier: Bachem Americas
Certificates
About this item
α-CGRP (8-37), a competitive CGRP1 receptor antagonist, exhibited hypothermic and antinociceptive effects. CGRP-I (8-37) antagonized the effects of amylin and amylin (1-8) on osteoblasts but was substantially less potent in this regard than amylin (8-37) (H-2746). Furthermore, it inhibited the evoked discharge frequency of wide dynamic range neurons in dorsal horn of the spinal cord in rats.
Specifications
- CAS No.:129121-73-9
- Species:RatMouse
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD18782272
- Protein Synonyms:Calcitonin Gene-Related Peptide I (8-37)|CGRP-I (8-37)
- Protein/Peptide Name:alpha-CGRP (8-37) Trifluoroacetate
- Molecular Weight:3127.56
- Sequence:H-Val-Thr-His-Arg-Leu-Ala-Gly-Leu-Leu-Ser-Arg-Ser-Gly-Gly-Val-Val-Lys-Asp-Asn-Phe-Val-Pro-Thr-Asn-Val-Gly-Ser-Glu-Ala-Phe-NH₂
- Molecular Formula:C₁₃₈H₂₂₄N₄₂O₄₁
Specifications
Frequently Bought Together