Dendroaspis Natriuretic Peptide
H-4904.0500BA
:
- CAS No.:255721-52-9
- Source:
- Species:
- Size:0.5 mg
- Environmentally Preferable:
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD02687290
- Protein Synonyms:DNP
- Protein/Peptide Name:Dendroaspis Natriuretic Peptide
- Molecular Weight:4190.69
- Sequence:H-Glu-Val-Lys-Tyr-Asp-Pro-Cys-Phe-Gly-His-Lys-Ile-Asp-Arg-Ile-Asn-His-Val-Ser-Asn-Leu-Gly-Cys-Pro-Ser-Leu-Arg-Asp-Pro-Arg-Pro-Asn-Ala-Pro-Ser-Thr-Ser-Ala-OH
- Molecular Formula:C₁₈₀H₂₈₂N₅₆O₅₆S₂
- Cat. No.:H-4904.0500BA
- Supplier no.:H-4904.0500
The 38 amino acid peptide DNP, which has been originally isolated from the venom of the green mamba snake (Dendroaspis angusticeps), shares structural homology with the family of natriuretic peptides. DNP contains a 17 amino acid disulfide ring similar to ANP, BNP, and CNP, which mediate biological actions through particulate guanylyl cyclase receptors and generation of guanosine monophosphate (cGMP).