Order Entry
United States
ContactUsLinkComponent
(D-Trp⁶,D-Leu⁷)-LHRH Trifluoroacetate
(D-Trp⁶,D-Leu⁷)-LHRH Trifluoroacetate
Supplier:  Bachem Americas
Certificates
(D-Trp⁶,D-Leu⁷)-LHRH Trifluoroacetate
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    321709-37-9
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD18633546
  • Protein Synonyms:
    (D-Leu7)-Triptorelin
  • Protein/Peptide Name:
    (D-Trp⁶,D-Leu⁷)-LHRH Trifluoroacetate
  • Molecular Weight:
    1311.47
  • Sequence:
    Pyr-His-Trp-Ser-Tyr-D-Trp-D-Leu-Arg-Pro-Gly-NH₂
  • Molecular Formula:
    C₆₄H₈₂N₁₈O₁₃

Specifications

Frequently Bought Together