H-Phe-Phe-Phe-Phe-Phe-OH
Supplier: Bachem Americas
Certificates
About this item
Pentaphenylalanine, FFFFF.
Specifications
- CAS No.:65757-10-0
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD00083786
- Protein Synonyms:pentaphenylalanine
- Protein/Peptide Name:H-Phe-Phe-Phe-Phe-Phe-OH
- Molecular Weight:753.90
- Sequence:H-Phe-Phe-Phe-Phe-Phe-OH
- Molecular Formula:C₄₅H₄₇N₅O₆
Specifications
Frequently Bought Together