(Des-Gly¹⁰,D-Ala⁶,Pro-NHEt⁹)-LHRH Acetate
Supplier: Bachem Americas
Certificates
About this item
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
Potent LHRH analog in rats and mice. Alarelin reversibly delays sexual maturation in rats. Applied as single drug or in combination with a dopamin receptor antagonist as domperidone for the stimulation and synchronization of ovulation and spawning in fish farming. CAS Number (net): 79561-22-1.
Specifications
- Source:
- Species:
- Environmentally Preferable:
Specifications
Frequently Bought Together