Human Melanocyte Protein PMEL 17 (130-138) Acetate
Catalog # H-3958.0001BA
Supplier: Avantor
CAS Number:
Specifications
- CAS No.:162558-08-9
- Species:Human
- Size:1 mg
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD01318803
- Protein Synonyms:Melanocyte Lineage-Specific Antigen GP100 (130-138)|Melanoma-Associated ME20 Antigen (130-138)|ME20-M/ME20-S (130-138)|ME20M/ME20S (130-138)|95 kDa Melanocyte-Specific Secreted Glycoprotein (130-138)
- Protein/Peptide Name:Melanocyte Protein PMEL 17 (130-138) Acetate
- Molecular Weight:1195.34
- Sequence:H-Lys-Thr-Trp-Gly-Gln-Tyr-Trp-Gln-Val-OH
- Molecular Formula:C₅₈H₇₈N₁₄O₁₄
- Cat. No.:H-3958.0001BA
- Supplier no.:H-3958.0001
Specifications
About this item
For further melanoma-associated peptides please see also Fibronectin Fragments/RGD Peptides, Melanin-Concentrating Hormones (MCH) and Related Peptides, Peptide 74, Thrombospondin-1 (TS1) Cell Binding Domain (CBD) Fragments.
PMEL 17 could be a melanogenic enzyme. This sequence has been identified as a gp100 epitope that is recognized in the context of HLA-A2 by tumor-infiltrating lymphocytes (TIL) in vivo.