Acetyl-(Nle⁴,Asp⁵,D-Phe⁷,Lys¹⁰)-cyclo-alpha-MSH (4-10) Amide Acetate
Melanotan II, a cyclic MSH analog, is a potent full agonist of the melanocortin-3 and melanocortin-4 receptors (MC3-R and MC4-R). Intracerebroventricular administration of MTII inhibited feeding in four different mouse models of hyperphagia. MTII is a valuable tool for the study of the agouti obesity syndrome and of the role of melanocortinergic neurons in feeding.
- CAS No.:121062-08-6
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD11045941
- Protein Synonyms:Melanotan II|MTII|Bremelanotide amide
- Protein/Peptide Name:Acetyl-(Nle⁴,Asp⁵,D-Phe⁷,Lys¹⁰)-cyclo-alpha-MSH (4-10) amide Acetate
- Molecular Weight:1024.19
- Sequence:Ac-Nle-cyclo(-Asp-His-D-Phe-Arg-Trp-Lys-NH₂)
- Molecular Formula:C₅₀H₆₉N₁₅O₉
Frequently Bought Together