Diprotin A
See also H-3974, H-Lys(4-nitro-Z)-pyrrolidide · HCl (N-1710) and H-Phe-pyrrolidide (E-3275).
An inhibitor of dipeptidyl aminopeptidase IV.
- CAS No.:90614-48-5
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD00038707
- Protein/Peptide Name:Diprotin A
- Molecular Weight:341.45
- Sequence:H-Ile-Pro-Ile-OH
- Molecular Formula:C₁₇H₃₁N₃O₄
Frequently Bought Together